Order Entry
Puerto Rico
ContactUsLinkComponent
Arsenic(III) oxide ≥99.95% ACS primary standard
Toxic
Health
Corrosive
Environment
Arsenic(III) oxide ≥99.95% ACS primary standard
Catalog # 100203-098
CAS Number:  
Toxic
Health
Corrosive
Environment
Arsenic(III) oxide ≥99.95% ACS primary standard
Catalog # 100203-098
Supplier Number:  33-3313-100G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 g
  • Cat. No.:
    100203-098
  • Supplier no.:
    33-3313.100

Specifications

About this item

 :
 :197.84 g/mol
Merck Index:12,00844
Boiling Pt:465 °C (subl.)
Melting Pt:315 °C
Density:3.738
Storage Temperature:Ambient
EINECS:215-481-4
UN:1561
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS05
 :GHS09
CAS Number:1327-53-3
Formula:As₂O₃
MDL Number:MFCD00003433