Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Copper(II) sulphide ≥99.5% (Cu basis)

Marketsource
Supplier:   Strem Chemicals Inc
Copper(II) sulphide ≥99.5% (Cu basis)
Marketsource
Certificates
Copper(II) sulphide ≥99.5% (Cu basis)

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Copper sulphide, Cupric sulphide, Copper(II)sulphide
  • CAS Number:1317-40-4
  • Formula:CuS
  • MDL Number:MFCD00016066
  • Molecular Weight:95.61 g/mol
  • Merck Index:12,02724
  • Density:4.6
  • EINECS:215-271-2
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    50 g
    Log in for availability
    Log in for availability
    Each (50g)
    Each (50g) - $120.31
    $120.31
    Each (50g)
       $120.31

    Supplier Number: 29-2969-50G
    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    250 g
    Log in for availability
    Log in for availability
    Each (250g)
    Each (250g) - $460.00
    $460.00
    Each (250g)
       $460.00

    Supplier Number: 29-2969-250G
    Specifications:
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.