Bromobenzene, Standard, 2000 μg/ml, Restek
RK30250
: Restek
:
- Pack type:Ampule
- CAS No.:108-86-1
- Size:1 ml
- Formula:C₆H₅Br
- Description:Bromobenzene
- Cat. No.:RK30250
- Supplier no.:30250
Solvent packaged in purge-and-trap-grade methanol.
- 2000 μg/mL in P&T methanol, 1 mL/ampoule
Packaged in purge-and-trap-grade methanol at 2000 μg/mL. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data Sheet.