Ethylbenzene-D10, Standard, 2000 μg/ml, Restek
Catalog # RK30029
Supplier: Restek
CAS Number:
Specifications
- Pack type:Ampule
- CAS No.:25837-05-2
- Size:1 ml
- Formula:C₇D₁₀
- Description:Ethylbenzene-D10
- Cat. No.:RK30029
- Supplier no.:30029
Specifications
About this item
Ethylbenzene-d10 is immersed in purge-and-trap-grade methanol.
- 2000 μg/mL in P&T methanol, 1 mL/ampoule
Packaged in purge-and-trap-grade methanol at 2000 μg/mL in P&T methanol. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data.