ContactUsLinkComponent
Semi-quantitative test strips QUANTOFIX Peroxide 100
Semi-quantitative test strips QUANTOFIX Peroxide 100
Catalog # 60787-736
Supplier:  Macherey-Nagel
CAS Number:  
undefined
Semi-quantitative test strips QUANTOFIX Peroxide 100
Catalog # 60787-736
Supplier:  Macherey-Nagel
Supplier Number:  91312
CAS Number:  
Specifications
  • Pack type:
    100 Sticks
  • Cat. No.:
    60787-736
  • Supplier no.:
    91312
Specifications
About this item
For semi-quantitative determinations
Gradation: 0 to 100mg/L. Color changes from white to blue.