ContactUsLinkComponent
Semi-quantitative test strips QUANTOFIX Sulfite
Semi-quantitative test strips QUANTOFIX Sulfite
Catalog # 60787-728
Supplier:  Macherey-Nagel
Small Business Enterprise
CAS Number:  
undefined
Semi-quantitative test strips QUANTOFIX Sulfite
Catalog # 60787-728
Supplier:  Macherey-Nagel
Supplier Number:  91306
Small Business Enterprise
CAS Number:  
Specifications
  • Pack type:
    100 Strips
  • Cat. No.:
    60787-728
  • Supplier no.:
    91306
Specifications
About this item
For semi-quantitative determinations
Gradation: 0 to 1000mg/L. Color changes from white to salmon.