IL-4 ELISA MAX™ Deluxe ELISA Kit, BioLegend
Catalog # 431104-BL
Supplier: Biolegend
CAS Number:
Specifications
- Conjugate ELISA:Unconjugated
- Primary antibody reactivity:Mouse
- Target protein:IL-4
- Description:IL-4 ELISA MAX™ Deluxe
- Size:5 Plates
- Cat. No.:431104-BL
- Supplier no.:431104
Specifications
About this item
IL-4 ELISA MAX™ Deluxe ELISA Kit reacts with Mouse