IL-2 ELISA MAX™ Standard ELISA Kit, BioLegend
431001-BL
:
- Conjugate ELISA:Unconjugated
- Primary antibody reactivity:Mouse
- Target protein:IL-2
- Description:IL-2 ELISA MAX™ Standard
- Size:5 Plates
- Cat. No.:431001-BL
- Supplier no.:431001
IL-2 ELISA MAX™ Standard ELISA Kit reacts with Mouse