TNF-alpha ELISA MAX™ Deluxe ELISA Kit, BioLegend
Catalog # 430204-BL
Supplier: Biolegend
CAS Number:
Specifications
- Conjugate ELISA:Unconjugated
- Primary antibody reactivity:Mouse
- Target protein:TNF-alpha
- Description:TNF-alpha ELISA MAX™ Deluxe
- Size:5 Plates
- Cat. No.:430204-BL
- Supplier no.:430204
Specifications
About this item
TNF-alpha ELISA MAX™ Deluxe ELISA Kit reacts with Human.