IL-5 MAX™ Deluxe ELISA Kit, BioLegend
Catalog # 430404-BL
Supplier: Biolegend
CAS Number:
Specifications
- Conjugate ELISA:Unconjugated
- Primary antibody reactivity:Human
- Target protein:IL-5
- Description:IL-5 ELISA MAX™ Deluxe
- Size:5 Plates
- Cat. No.:430404-BL
- Supplier no.:430404
Specifications
About this item
IL-5 ELISA MAX™ Deluxe ELISA Kit reacts with Human