Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 95058-120
Supplier:  KPL
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 95058-120
Supplier:  KPL
Supplier Number:  5230-0297

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Pig
  • Western Blot:
    Yes
  • Size:
    500 µg
  • Epitope:
    Heavy and Light chain
  • Cat. No.:
    95058-120
  • Supplier no.:
    02-14-06

Specifications

About this item

Secondary Goat Anti-IgG (H+L) Reacts with Pig (Swine, Porcine)

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Pig