Order Entry
United States
ContactUsLinkComponent
Anti-Deer IgG Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-Deer IgG Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 95058-184
Supplier:  KPL
Anti-Deer IgG Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 95058-184
Supplier:  KPL
Supplier Number:  5230-0329
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Deer
  • Western Blot:
    Yes
  • Size:
    500 µg
  • Environmentally Preferable:
  • Epitope:
    Heavy and Light chain
  • Cat. No.:
    95058-184
  • Supplier no.:
    02-31-06

Specifications

About this item

Secondary Rabbit Anti-IgG (H+L) Reacts with Deer

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Deer