Order Entry
Puerto Rico
ContactUsLinkComponent
Formaldehyde 16% in aqueous solution, Ultrapure methanol-free
Toxic
Corrosive
Health
Formaldehyde 16% in aqueous solution, Ultrapure methanol-free
Catalog # 87001-904
Supplier:  Polysciences Inc
Product available on GSA Advantage®
Small Business Enterprise
CAS Number:  
undefined
Toxic
Corrosive
Health
Formaldehyde 16% in aqueous solution, Ultrapure methanol-free
Catalog # 87001-904
Supplier:  Polysciences Inc
Supplier Number:  18814-20
Product available on GSA Advantage®
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    10 ml
  • Cat. No.:
    87001-904
  • Supplier no.:
    18814-20

Specifications

About this item

 :
 :30.03 g/mol
Merck Index:13,04259
Boiling Pt:–19.3 °C (1013 hPa)
Melting Pt:–92 °C
Density:0.8153 g/cm³ (–20 °C)
Storage Temperature:Ambient
EINECS:200-001-8
UN:3334
ADR:9,
 :GHS06
 :GHS05
 :GHS08
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274