Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
VWR® TBS / TWEEN® 20 Buffer

VWR® TBS / TWEEN® 20 Buffer

Supplier:   VWR
VWR® TBS / TWEEN® 20 Buffer
Supplier:  VWR
Certificates
VWR® TBS / TWEEN® 20 Buffer
VWR® TBS / TWEEN® 20 Buffer

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

VWR® offers a comprehensive line of buffers and detergents ideal for use in applications such as Western blotting and ELISA.

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 500 mL
    Discontinued
    Supplier Number: -
  • 10791-792
    TBS / TWEEN® 20 Buffer, 20X
    TBS / TWEEN® 20 Buffer, 20X
    1 L
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $317.78
    $317.78
    Each (1l)
       $317.78

    Supplier Number: K873-1L
  • 10791-794
    TBS / TWEEN® 20 Buffer, 20X
    TBS / TWEEN® 20 Buffer, 20X
    4 L
    Log in for availability
    Log in for availability
    Each (4l)
    Each (4l) - $513.97
    $513.97
    Each (4l)
       $513.97

    Supplier Number: K873-4L
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.