A800 Series Eyewear, Sperian Eye and Face Protection, ORS Nasco
Catalog # 300053-408
Supplier: Avantor
Specifications
- Frame color:Gray
- Description:Wrap-around lens safety glasses
- Environmentally Preferable:
- Cat. No.:300053-408
- Lens coating:Scratch-resistant/hardcoat
- Lens tint:Blue mirror
- Supplier no.:812-A803
- Lens material:PC
Specifications
About this item
These glasses feature soft padded inserts which run down the length of the temple.
- Wrap-aroud style
- Impact- and scratch-resistant
- Soft nosepiece for added comfort
Certifications: Complies with ANSI Z87+ High Impact Standards.