jetPEI®-Hepatocyte, primary hepatocytes transfection reagent
89129-944
: Avantor
:
- Cell Specific:Hepatocyte
- DNA:Yes
- Description:jetPEI®-Hepatocyte, DNA transfection reagent
- Size:500 µl
- Environmentally Preferable:
- Cat. No.:89129-944
- Buffer volume:50 ml
- No. of reactions:160 (1 µg of DNA per well, 24 wells)
- Supplier no.:102-05N
jetPEI®-Hepatocyte is a DNA transfection reagent designed to transfect Hepatocyte-like cells.
- Efficient transfection of primary hepatocytes (up to 50% of transfection efficiency in hepatocyte-like cells and primary hepatocytes)
- Gentle to the cells
- Easy-to-use protocol
- Compatible with serum and antibiotics
jetPEI®-Hepatocyte is recommended to transfect primary hepatocytes and cell lines such as human hepatocarcinoma HepG2 and primary hepatocytes.
: Since 2002, the company is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Every batch of jetPEI®-Hepatocyte is tested in a transfection assay. Typically, transfection of a firefly luciferase gene under the control of the CMV promoter gives 109 RLU (relative light unit)/mg of protein. The value for each batch is indicated on the Certificate of Analysis.
Every batch of jetPEI®-Hepatocyte is tested in a transfection assay. Typically, transfection of a firefly luciferase gene under the control of the CMV promoter gives 109 RLU (relative light unit)/mg of protein. The value for each batch is indicated on the Certificate of Analysis.
: 150 mM NaCl solution is included. 0.5 ml of jetPEI®-Hepatocyte transfection reagent is sufficient to perform up to 160 transfections in 24-well plates.