Order Entry
Puerto Rico
ContactUsLinkComponent
Hematoxylin solution Gill II for histology, for cytology
Hematoxylin solution Gill II for histology, for cytology
Catalog # 87001-922
Supplier:  Polysciences Inc
Product available on GSA Advantage®
Small Business Enterprise
CAS Number:  
undefined
Hematoxylin solution Gill II for histology, for cytology
Catalog # 87001-922
Supplier:  Polysciences Inc
Supplier Number:  24243-500
Product available on GSA Advantage®
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 ml
  • Cat. No.:
    87001-922
  • Supplier no.:
    24243-500

Specifications

About this item

 :
 :302.28 g/mol
Storage Temperature:Ambient
CAS Number:517-28-2
 :208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111