ContactUsLinkComponent
2D-Xtract™, G-Biosciences
2D-Xtract™, G-Biosciences
  95043-430
 :  
undefined
2D-Xtract™, G-Biosciences
  95043-430
 :  786-501
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    2D-Xtract™
  • Size:
    50 ml
  • Cat. No.:
    95043-430
  • Supplier no.:
    786-501
2D-Xtract™ is a dry, urea-based and ready-to-use protein solubilization buffer.
  • Convenient and simple to use extraction buffer
  • No preparation required, simply hydrate and use
  • Prevents urea induced protein carbamylation
  • Prevents waste of unused reagents
  • No artifactual protein bands due to dust and human skin contamination
2D-Xtract™ is a proprietary modification of urea based chaotropic extraction buffers. Normal urea based buffers solubilize proteins effectively however can modify the native charge of the proteins, due to carbamylation. A process that modifies amino groups, inducing changes in the isoelectric point of proteinsleading to artifactual 2D results.
2D-Xtract™ is a dry, pre-mixed formulation of urea, thiourea, CHAPS and non detergent sulfobetaine (ND SB 201) for maximumsolubilizing strength. The dry format allows researchers to freshly rehydrate as much or as little as is required and therefore prevent urea induced carbamylation.
This is suitable for sample extraction and solubilization for 2D gel electrophoresis and other applications and also suitable for rehydration of IPG Strips.
 : Accessories are available.
 : The kit is shipped at ambient temperature. Upon arrival, store it as specified on the bottle labels.