ContactUsLinkComponent
TearTech® 25 Holes Foam Rack, Therapak®
TearTech® 25 Holes Foam Rack, Therapak®
  89171-120
undefined
TearTech® 25 Holes Foam Rack, Therapak®
  89171-120
 :  68825
  • Description:
    Foam rack
  • Cat. No.:
    89171-120
  • Supplier no.:
    68825
When combined with shipping boxes, specimen bags or insulated shippers, this foam tube rack provides cushioning and protection for 13x75mm tubes.
The unique die-cut construction allows for tearing away variable-sized grids to customize transport load. Overall foam block size is 150x150x100mm