Haematoxylin modified solution (Harris)
Product Details & Documents
This regressive nuclear stain is much like Mayer’s Hematoxylin but is more stable.
- No Mercury
- Ready to Use Formula
- Exceptional Nuclear Detail
The formulation does not contain mercury.
This solution is ready to use and provides exceptional nuclear detail when used in progressive or regressive protocols.
- CAS Number:517-28-2
- Formula:C₁₆H₁₄O₆
- MDL Number:MFCD00078111
- Molecular Weight:302.28 g/mol
Documents
Specifications
Specifications
- Size:3.8 L (1 gal.)
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



