MAS™ CardioImmune™ XL Controls, Thermo Scientific
Supplier: Microgenics Corporation (Mas)
Certificates
About this item
Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte MAS™ CardioImmune™ XL Controls
Human serum matrix; Offers 4 distinctive levels; Assayed values available for most major analyzers; Available in a tri-level multipack.
Analytes: BNP 32, Myoglobin, CK-MB, NT-pro BNP, Digitoxin, Troponin I, Homocysteine, Troponin T, hsCRP.
These controls are also available in a convenient Vista™ vial format.
Ordering information: Available in Vista™ vial format. Designed to monitor serum cardiac marker test methods.