BIO-BAG™ Environmental Chambers, BD™
90003-711
: BD
- Description:Bio-Bag™ type C, anaerobic transport bag
- Cat. No.:90003-711
- Supplier no.:261510
Incorporate traditional gas-generation methods into a reliable pouch system that does not require the addition of water.
The type A multi-plate bag is for the isolation of anaerobic bacteria, and is sized to hold three standard Petri dishes, identification strips, or MIC plates. It comes with 50 anaerobic generators/indicators and 100 Palladium catalyst containers. The Type Cfj bag allows C. jejuni cultures to be set up and observed for growth individually, so that appropriate atmospheric conditions are not interrupted. It is sized to hold one or two petri dishes, and comes with 100 microaerophilic generators.