4-CORE® Buffer Pack, Promega®
4-CORE® Buffer Pack, 4 x 1 mL
Supplier: Promega Corporation
Supplier #: R9921
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Buffer pack contains convenient aliquots of Promega® restriction enzyme 10X buffers A, B, C, and D
The majority of Promega® restriction enzymes have optimal activity in one of these four 10X reaction buffers.
Ordering information: Pack contains 1 mL each of buffers A, B, C, and D.
Caution: Note: For laboratory use. Outside of the United States, this product is intended for research use only, unless otherwise stated.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:4-CORE® Buffer Pack
- Size:4 x 1 mL
- Cat. No.:PAR9921
- Storage Temperature:-30 to -10°C



