ContactUsLinkComponent
Hanging Dual Scale Thermometer, Thermco
Hanging Dual Scale Thermometer, Thermco
Catalog # 89168-870
Supplier:  Thermco
undefined
Hanging Dual Scale Thermometer, Thermco
Catalog # 89168-870
Supplier:  Thermco
Supplier Number:  BFZ80
Specifications
  • Cat. No.:
    89168-870
  • Range:
    −40…+25 °C (−40…+80 °F)
  • Supplier no.:
    BFZ80
Specifications
About this item
This non-mercury hanging thermometer offers a dual scale of Celsius and Fahrenheit
It works for both refrigerators and freezers, and the tube is completely enclosed in plastic for safety. Includes stainless steel shelf hangers with keyhole backs for wall hanging