Orders LinkContactUsLinkComponent
Urinometer, Profex Medical Products
Urinometer for determining the specific gravity of urine.
  15744-325
 :  760
Urinometer, Profex Medical Products
  15744-325
Urinometer, Profex Medical Products
Urinometer for determining the specific gravity of urine.
$149.95
Each Case of 12
Glass urinometer has a reinforced, clear, heavy fluted jar with extra-wide base and pouring tip
  • Specific gravity range: 1.000 to 1.060
Shot-weighted hydrometer float.
  • Description:
    Urinometer for determining the specific gravity of urine.