

Tetrahydrofuran-[D₈] 99.5 atom % D for NMR spectroscopy
Catalog # 200007-840
Supplier: Avantor
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Ampoule
- Size:7.5 ml (10×0.75 ml)
- Cat. No.:200007-840
- Supplier no.:32078-0075|AC32078-0075
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.05 % (Coulometric)
- D-Enrichment:≥99.4 %
Specification Test Results
About this item
:
:80.04 g/mol
Boiling Pt:65 °C (1013 hPa)
Melting Pt:–106 °C
Density:0.98 g/cm³
Flash Pt:–21 °C (closed cup)
Storage Temperature:Refrigerator
EINECS:216-898-4
UN:2056
ADR:3,II
:GHS02
:GHS07
CAS Number:1693-74-9
Formula:C₄D₈O
MDL Number:MFCD00044238