Order Entry
Puerto Rico
ContactUsLinkComponent
 
Health
Irritant
Manganese(IV) oxide for synthesis, (precipitated active), Sigma-Aldrich®
Catalog # EM8.05958.1000
Supplier:  MilliporeSigma
CAS Number:  
Health
Irritant
Manganese(IV) oxide for synthesis, (precipitated active), Sigma-Aldrich®
Catalog # EM8.05958.1000
Supplier:  MilliporeSigma
Supplier Number:  8.05958.1000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 kg
  • Cat. No.:
    EM8.05958.1000
  • Supplier no.:
    8.05958.1000

Specifications

About this item

 :
 :86.94 g/mol
Merck Index:14,05730
Melting Pt:535 °C
Density:5.03 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
EINECS:215-202-6
 :GHS08
 :GHS07
CAS Number:1313-13-9
Formula:MnO₂
MDL Number:MFCD00003463