Order Entry
Puerto Rico
ContactUsLinkComponent
Acetone ≥99.5% (by GC), Finyte® for microelectronic, J.T.Baker®
Flammable
Irritant
Health
Acetone ≥99.5% (by GC), Finyte® for microelectronic, J.T.Baker®
Catalog # JT5008-5
CAS Number:  
undefined
Flammable
Irritant
Health
Acetone ≥99.5% (by GC), Finyte® for microelectronic, J.T.Baker®
Catalog # JT5008-5
Supplier Number:  5008-05
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly bottle, pressure bottom
  • Size:
    4 L
  • Cat. No.:
    JT5008-5
  • Supplier no.:
    5008-05

Specifications

Specification Test Results

  • Assay (by GC):
    ≥99.5 %
  • Color (APHA):
    ≤10
  • Acidity (µeq/g):
    ≤0.3
  • Aldehydes (as HCHO):
    ≤20000 ppb
  • Alkalinity (µeq/g):
    ≤0.5
  • Isopropyl Alcohol (CH₃CHOHCH₃)(BY WT):
    ≤0.05 %
  • Methanol (CH₃OH)(BY WT):
    ≤0.05 %
  • Residue after Evaporation:
    ≤3000.0 ppb
  • Water (H₂O):
    ≤0.50 %
  • Solubility in H₂O:
    Passes Test
  • Chloride (Cl):
    ≤200.0 ppb
  • Phosphate (PO₄):
    ≤50.0 ppb
  • Trace Impurities - Aluminum (Al):
    ≤10.0 ppb
  • Trace Impurities - Antimony (Sb):
    ≤5.0 ppb
  • Trace Impurities - Arsenic (As):
    ≤5.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤10.0 ppb
  • Trace Impurities - Beryllium (Be):
    ≤10.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤10.0 ppb
  • Trace Impurities - Boron (B):
    ≤10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤10.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤10.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤10.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤10.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤10.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤10.0 ppb
  • Trace Impurities - Gold (Au):
    ≤10.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤10.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤10.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤10.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤10.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤10.0 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤10.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤10.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤10.0 ppb
  • Trace Impurities - Silicon:
    ≤10.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤10.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤10.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤10.0 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤10.0 ppb
  • Trace Impurities - Thallium (Tl):
    ≤10.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤10.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤10.0 ppb
  • Trace Impurities - Vanadium (V):
    ≤10.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤10.0 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤10.0 ppb
  • Particle Count - 0.5 µm and greater:
    ≤50 par/ml
  • Particle Count - 1.0 µm and greater:
    ≤8 par/ml

Specification Test Results

About this item

 :
 :58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
 :Management of Change (MOC) category = R
REACH:01-2119471330-49
 :GHS02
 :GHS07
 :GHS08
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765