



Sodium methoxide ∼30% (w/w) in methanol
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:124-41-4
:
Formula:CH₃NaO
MDL Number:MFCD00012179
:54.02 g/mol
Merck Index:14,8643
Boiling Pt:93 °C
Melting Pt:<6 °C
Density:0.969
Flash Pt:16 °C(61 °F)
EINECS:204-699-5
UN:1289
ADR:3,III
:GHS02
:GHS05
:GHS06
:GHS08
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
VWR GHS Compliance Letter (US)