Valeric acid 99%
About this item
Purity: 99% CAS No.: 109-52-4 Molecular Formula: C5H10O2 Formula Weight: 102.13 Flash Point: 88°(190°F) Refractive Index: 1.4080 Odor: Unpleasant Melting Point: -34° Boiling Point: 184-186° Beilstein Reference: 969454 Merck Reference: 14,9904 UN No.: 3265 MDL No.: MFCD00004413
- CAS Number:109-52-4
- Formula:C₅H₁₀O₂
- MDL Number:MFCD00004413
- Molecular Weight:102.13 g/mol
- Merck Index:14,9904
- Boiling Pt:184…186 °C
- Melting Pt:–34 °C
- Density:0.938
- Flash Pt:88 °C (190 °F)
- Storage Temperature:Ambient
- EINECS:203-677-2
- UN:3265
- ADR:8,III
2 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 109-52-4 Molecular Formula: C5H10O2 Formula Weight: 102.13 Flash Point: 88°(190°F) Refractive Index: 1.4080 Odor: Unpleasant Melting Point: -34° Boiling Point: 184-186° Beilstein Reference: 969454 Merck Reference: 14,9904 UN No.: 3265 MDL No.: MFCD00004413
- CAS Number:109-52-4
- Formula:C₅H₁₀O₂
- MDL Number:MFCD00004413
- Molecular Weight:102.13 g/mol
- Merck Index:14,9904
- Boiling Pt:184…186 °C
- Melting Pt:–34 °C
- Density:0.938
- Flash Pt:88 °C (190 °F)
- Storage Temperature:Ambient
- EINECS:203-677-2
- UN:3265
- ADR:8,III
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



