

Tetrahydrofuran-[D₈] (100% D) (Isotopic) for NMR spectroscopy
Catalog # AA42348-KV
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Pack type:Bottle
- Size:10 each
- Cat. No.:AA42348-KV
- Supplier no.:42348-KV
Specifications
About this item
CAS Number:1693-74-9
:
Formula:C₄D₈O
MDL Number:MFCD00044238
:80.04 g/mol
Boiling Pt:66 °C
Boiling Pt:65 °C (1013 hPa)
Melting Pt:–106 °C
Melting Pt:-108 °C
Density:0.98 g/cm³ (20 °C)
Flash Pt:–17 °C (1 °F)
Flash Pt:-17 °C (closed cup)
Storage Temperature:Ambient
EINECS:216-898-4
UN:2056
ADR:3,II
:GHS02
:GHS07