


Methanol-[D₄] (99.8% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:811-98-3
:
Formula:D₃COD
MDL Number:MFCD00044637
:36.01 g/mol
Boiling Pt:64.7 °C
Melting Pt:–98 °C
Density:0.89
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:212-378-6
UN:1230
ADR:3,II
:GHS02
:GHS06
:GHS08
Related Information
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
VWR GHS Compliance Letter (US)