Order Entry
Puerto Rico
ContactUsLinkComponent
1,2-Dichlorobenzene-[D₄] (99% D) (Isotopic) for NMR spectroscopy
Irritant
1,2-Dichlorobenzene-[D₄] (99% D) (Isotopic) for NMR spectroscopy
Catalog # AA42280-06
CAS Number:  
Irritant
1,2-Dichlorobenzene-[D₄] (99% D) (Isotopic) for NMR spectroscopy
Catalog # AA42280-06
Supplier Number:  042280-06
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Size:
    5 g
  • Cat. No.:
    AA42280-06
  • Supplier no.:
    42280-06

Specifications

About this item

CAS Number:2199-69-1
 :
Formula:C₆D₄Cl₂
MDL Number:MFCD00037106
Boiling Pt:181 °C
Melting Pt:–17 °C
Density:1.34
Flash Pt:65 °C (150 °F)
EINECS:218-606-0
ADR:6.1,III
 :150.97 g/mol
UN:1591
 :GHS07
Storage Temperature:Ambient