Tetrahydrofuran ≥99.7% stabilised, non-UV for HPLC
: Butylene oxide, THF, Diethylene oxide
:109-99-9
:
:C₄H₈O
:MFCD00005356
:66
:0.889
:250 ppm Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
:72.11 g/mol
:-108.5
:–17 °C (1 °F)
:2056
:3 II
:14,9211
:Ambient
:203-726-8
:05-2114615188-48
:GHS02
:GHS08
:GHS07
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.