Order Entry
Puerto Rico
ContactUsLinkComponent
Zinc selenide, crystal optic disc 13 mm x 2 mm, polished both sides
Toxic
Health
Environment
Zinc selenide, crystal optic disc 13 mm x 2 mm, polished both sides
Catalog # AA38438-KT
CAS Number:  
Toxic
Health
Environment
Zinc selenide, crystal optic disc 13 mm x 2 mm, polished both sides
Catalog # AA38438-KT
Supplier Number:  038438-KT
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    1 each
  • Cat. No.:
    AA38438-KT
  • Supplier no.:
    38438-KT

Specifications

About this item

CAS Number:1315-09-9
 :
Formula:SeZn
MDL Number:MFCD00011296
 :144.35 g/mol
Merck Index:12,10290
Melting Pt:1793 °C
Density:5.4 g/cm³ (20 °C)
EINECS:215-259-7
UN:3283
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS09
Storage Temperature:Ambient