Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Silver nitrate standardised solution 0.1 N in aqueous solution

Corrosive
Environment
Silver nitrate standardised solution 0.1 N in aqueous solution
Certificates
Silver nitrate standardised solution 0.1 N in aqueous solution
Silver nitrate standardised solution 0.1 N in aqueous solution

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00003414

CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Colorless Odor: Odorless UN No.: 3082 MDL No.: MFCD00003414

Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:3082
  • ADR:9,III
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $252.26
    $252.26
    Each (500ml)
       $252.26

    Supplier Number: 035601-AP
  • Drum
    20 L
    Log in for availability
    Log in for availability
    Each (20l)
    Each (20l) - $2,984.14
    $2,984.14
    Each (20l)
       $2,984.14

    Supplier Number: 035601-M4

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.