Order Entry
Puerto Rico
ContactUsLinkComponent
Clear Bath® Algae Inhibitor, Spectrum Labs
Clear Bath® Algae Inhibitor, Spectrum Labs
Catalog # 54847-540
Supplier:  Avantor
Small Business Enterprise
CAS Number:  
Clear Bath® Algae Inhibitor, Spectrum Labs
Catalog # 54847-540
Supplier:  Avantor
Small Business Enterprise
CAS Number:  

Specifications

  • Size:
    237 mL (8 oz.)
  • Cat. No.:
    54847-540
  • Supplier no.:
    105540

Specifications

About this item

This solution keeps water baths and other equipment clean and algae-free.

It does not contain toxic copper compounds or other heavy metals that can accumulate in groundwater after disposal. The active ingredient is an organic compound that inhibits algae growth without impacting the environment. Only 3 drops per liter of water (12 drops per gallon) are required, so one bottle can treat over 1500L (400gal.) of water.

Ordering information: EPA regulations do not allow for this product to be shipped in CA.