
Benzamidine hydrochloride hydrate ≥98.0% (by anhydrous basis), Proteomics Grade
97063-856
: VWR
:
- Pack type:HDPE bottle
- Size:100 g
- Cat. No.:97063-856
- Supplier no.:M141-100G
- Em (Lambda Max, MEOH, anhy):9500
- Heavy Metals (as Pb):0.005 %
- Identification (IR):PASS
- Melting Point:77 - 87 °C
- Purity (anhydrous):98.0 %
- Reassay Date:REPORT
- Solubility (1%, Water):NONE
:
:174.63 g/mol
:216-795-4
:GHS07
:Ambient
:206752-36-5
:1670-14-0
:C₆H₅C(=NH)NH₂·HCl·H₂O
:MFCD00066285