Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
VWR® dTTP

VWR® dTTP

dTTP 100 mM solution, pH 7.0, 40 µmoles
Catalog # 97062-872
Supplier:   VWR
Supplier #:  E542-40UM
VWR® dTTP
Catalog # 97062-872
Supplier:  VWR
CAS Number:  
VWR® dTTP
VWR® dTTP

dTTP 100 mM solution, pH 7.0, 40 µmoles

CAS Number:  
Price
$159.56
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Deoxyribonucleotides are the foundation of DNA and it takes thousands of these nucleotides to make up DNA. DNA consists of a sugar molecule (deoxyribose), a nitrogenous base (purine in adenine and guanine, pyrimidine in thymine or cytosine) and a phosphate molecule. VWR® deoxyribonucleotides are ideal for the amplification of DNA.

  • Building blocks for the amplification of DNA
  • High purity grade
Specifications

Specifications

  • Description:
    dTTP 100 mM solution, pH 7.0
  • Size:
    40 µmoles
  • Storage Conditions:
    -20°C
  • Cat. No.:
    97062-872
  • pH:
    7.0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.