Agarose, LF™ for pulsed field gel electrophoresis (PFGE)
Supplier: VWR
Certificates
About this item
CAS Number:9012-36-6
:
MDL Number:MFCD00081294
Boiling Pt:1413 °C
Melting Pt:801 °C
Storage Temperature:Ambient
EINECS:232-731-8
Specification Test Results
- DNase:none detected
- EEO (Electroendosmosis):< = 0.06
- Endonuclease/ligase inhibitory factors:none
- Gel Strength:> = 2000 g/cm2
- Gelling Range:37°C - 41°C
- Melting Range:93°C - 96°C
- Moisture:< = 8.5 %
- Protease:none detected
- RNase:none detected
- Sulfate:< = 0.06 %
Specification Test Results
Frequently Bought Together