VWR® SM Buffer, Ultra Pure Grade
Catalog # 97063-290
Supplier: VWR
CAS Number:
Specifications
- Description:SM BUFFER
- Size:500 mL
- Cat. No.:97063-290
- pH:7.40 - 7.60
- Supplier no.:J614-500ML
Specifications
About this item
- Clear haze-free solution
- DNase-free