Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Cloning Buffers
VWR® Sodium Acetate, 3M Solution, pH 5.2, Biotechnology Grade
VWR® Sodium Acetate, 3M Solution, pH 5.2, Biotechnology Grade
VWR® Sodium Acetate, 3M Solution, pH 5.2, Biotechnology Grade
Catalog # 97062-834
Supplier:
VWR
CAS Number:
SDS
Certificates
Expand image
VWR® Sodium Acetate, 3M Solution, pH 5.2, Biotechnology Grade
Catalog # 97062-834
Supplier:
VWR
Supplier Number:
E521-100ML
CAS Number:
SDS
Certificates
Specifications
Description:
Sodium acetate solution 3M, pH 5.2
Size:
100 ml
Cat. No.:
97062-834
Supplier no.:
E521-100ML
Specifications
About this item
Product Details
Documentation
Product Details
Documentation