Linoleic acid, Ultrapure
Catalog # 97062-128
Supplier: VWR
CAS Number:
Specifications
- Size:10 g
- Cat. No.:97062-128
- Supplier no.:0660-10G
Specifications
Specification Test Results
- Purity:>= 99 %
Specification Test Results
About this item
:
:280.45 g/mol
Melting Pt:–5 °C
CAS Number:60-33-3
Formula:C₁₈H₃₂O₂
MDL Number:MFCD00064241
Melting Pt:-5°C