

Ammonium peroxodisulphate (APS) ≥98%, Pro-Pure, Proteomics Grade
Supplier: VWR
Certificates
About this item
CAS Number:7727-54-0
:
Formula:(NH₄)₂S₂O₈
MDL Number:MFCD00003390
:228.2 g/mol
Merck Index:13,00545
Melting Pt:120 °C
Density:1.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-786-5
UN:1444
ADR:5.1,III
:GHS07
:GHS08
Specification Test Results
- Chloride & Chlorate:<= 0.001%
- DNase:none detected
- Electrophoresis (P/F):Pass
- Heavy Metals (as Pb):<= 0.005%
- Insolubles:<= 0.005%
- Iron:<= 0.001%
- Manganese:<= 0.00005%
- Protease:none detected
- Purity:>= 98%
- Residue on Ignition:<= 0.05%
- RNase:none detected
- Titrable Free Acid:<= 0.04 meq/g
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Top Rated Guide
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together