Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Silver nitrate, Proteomics Grade

Silver nitrate, Proteomics Grade

Irritant
Health
Environment
Supplier:   VWR
Silver nitrate, Proteomics Grade
Supplier:  VWR
Certificates
Silver nitrate, Proteomics Grade

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Merck Index:12,08661
  • Boiling Pt:440 °C
  • Melting Pt:212 °C
  • Density:4.35 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II
  • REACH:01-2119513705-43
Specifications for Product Family

Specification Test Results

  • Chloride:
    < 0.0005%
  • Clarity of Solution:
    Pass
  • Copper:
    <= 0.0002 %
  • Free Acid Test:
    Pass
  • Iron:
    < = 0.0002 %
  • Lead:
    < = 0.001 %
  • Purity:
    > = 99 %
  • Substances not Precipitated by HCl:
    < 0.01 %
  • Sulfate:
    <= 0.002 %
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle, amber
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $775.92
    $775.92
    Each (100g)
       $775.92

    Supplier Number: M122-100G
  • Bottle, amber
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $2,847.48
    $2,847.48
    Each (500g)
       $2,847.48

    Supplier Number: M122-500G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.