
Guanidinium hydrochloride, Technical Grade low ash residue after ignition
Supplier: VWR
Certificates
About this item
CAS Number:50-01-1
:
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026
:95.53 g/mol
Merck Index:13,04578
Melting Pt:185 °C
Density:1.354 g/cm³ (20 °C)
EINECS:200-002-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Ammonium Chloride:< 0.5%
- Ash:< 0.4 %
- Moisture (KF):< 0.3 %
- Purity:> 98.5 %
Specification Test Results
Frequently Bought Together