Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium azide, high purity
Toxic
Environment
Sodium azide, high purity
Catalog # 97064-646
Supplier:  VWR
CAS Number:  
undefined
Toxic
Environment
Sodium azide, high purity
Catalog # 97064-646
Supplier:  VWR
Supplier Number:  0639-250G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    HDPE bottle
  • Size:
    250 g
  • Cat. No.:
    97064-646
  • Supplier no.:
    0639-250G

Specifications

About this item

 :
 :65.01 g/mol
Merck Index:13,08653
Melting Pt:275 °C
Density:1.85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:247-852-1
UN:1687
ADR:6.1,II
 :GHS06
 :GHS09
CAS Number:26628-22-8
Formula:NaN₃
MDL Number:MFCD00003536