Order Entry
Puerto Rico
ContactUsLinkComponent
Sealed Air Bubble Wrap® Cold Seal AirCap® Cohesive Coated Air Cellular Cushioning Material
Sealed Air Bubble Wrap® Cold Seal AirCap® Cohesive Coated Air Cellular Cushioning Material
Catalog # 500007-341
undefined
Sealed Air Bubble Wrap® Cold Seal AirCap® Cohesive Coated Air Cellular Cushioning Material
Catalog # 500007-341
Supplier Number:  SEL69566

Specifications

  • Description:
    Packaging with cohesive (self-clinging) bubble, with perforation for every 12"
  • Length:
    175'
  • Thickness:
    0.5 cm (0.2")
  • Cat. No.:
    500007-341
  • Width:
    12"
  • Supplier no.:
    SEL69566

Specifications

About this item

Self-cling, high-performance cushioning requires no tape.

  • Provides better protection
  • Uses less packaging material compared to loose fill or paper
  • Does not stick to product being protected

Barrier Bubble® technology allows to simply wrap item and press cohesive surfaces together.