Order Entry
Puerto Rico
ContactUsLinkComponent
PurSwab® Flexible Paddle Foam Tipped Applicator, Solid Polypropylene Handle, Puritan Medical Products
PurSwab® Flexible Paddle Foam Tipped Applicator, Solid Polypropylene Handle, Puritan Medical Products
  89133-780
Product available on GSA Advantage®
undefined
PurSwab® Flexible Paddle Foam Tipped Applicator, Solid Polypropylene Handle, Puritan Medical Products
  89133-780
 :  1802-PFSOLID
Product available on GSA Advantage®

 

  • Cat. No.:
    89133-780
  • Handle length:
    60.3 mm (2.374")
  • Head width:
    4.77 mm (0.188")
  • Head length:
    15.1 mm (0.594")
  • Handle width:
    2.54 mm (0.1")
  • Supplier no.:
    1802-PF SOLID

 

 

General-use swab for cleaning in critical environments and removing contamination and residues

Made in the USA.

Solid polypropylene shaft. Flexible paddle tip. Soft non-abrasive head. Lint and residue free. Nonsterile.

 : CE marked.
 : Packaged 50 per pack, 20 packs per case.