KIMTECH SCIENCE® Bench Top Protector Sheets, Kimberly-Clark Professional®
Catalog # 94001-646
Supplier: Avantor
Specifications
- Environmentally Preferable:
- Cat. No.:94001-646
- W×L:45.7 x 49.7 cm (18 x 191/2")
- Supplier no.:75460
Specifications
About this item
This advanced bench top protector features a polyethylene film backing that provides excellent slip resistance, creates an effective fluid barrier, and resists harsh chemicals
The white cellulose fiber top layer provides an absorbent, stable surface and protects experiments and delicate glassware. The bench top protector remains flat even after drying.